SLW4546/32
Pyrex® Spare sealing disc for high temperature screwcaps
natural, GL 32
Synonym(s):
Pyrex, SciLabware 4546/32
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NB.17
material
PTFE face
natural
manufacturer/tradename
4546/32
SciLabware
fitting
GL 32
Looking for similar products? Visit Product Comparison Guide
General description
For use with SLW4526 screwcaps.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service