GF43765937
Gold
mesh, nominal aperture 0.011 mm, size 10 × 10 mm, thickness 0.004 mm
Synonym(s):
AU008721
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
mesh
manufacturer/tradename
Goodfellow 437-659-37
resistivity
2.05 μΩ-cm, 0°C
nominal aperture
0.011 mm
size
10 × 10 mm
thickness
0.004 mm
bp
2808 °C (lit.)
mp
1063 °C (lit.)
density
19.3 g/mL at 25 °C (lit.)
SMILES string
[Au]
InChI
1S/Au
InChI key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service