Skip to Content
Merck
All Photos(1)

Key Documents

677043

Sigma-Aldrich

(2R,5R)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane

Synonym(s):

R,R-Me-RajPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21O2P
CAS Number:
Molecular Weight:
264.30
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

optical activity

[α]22/D -38.0°, c = 1 in chloroform

refractive index

n20/D 1.572

density

1.1052 g/mL at 25 °C

SMILES string

C[C@@H](P1C2=C(C=CC=C2)C3OCCO3)CC[C@H]1C

InChI

1S/C15H21O2P/c1-11-7-8-12(2)18(11)14-6-4-3-5-13(14)15-16-9-10-17-15/h3-6,11-12,15H,7-10H2,1-2H3/t11-,12-/m1/s1

InChI key

JAKXTLMNIHPLEU-VXGBXAGGSA-N

Application

DuPhos and BPE Ligands: Highly Efficient Privileged Ligands

  • Catalytic ligand used in copper-catalyzed addition reactions
Kanata purity

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.
RajPhos is a trademark of Kanata Chemical Technologies, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service