Skip to Content
Merck
All Photos(1)

Documents

674893

Sigma-Aldrich

3,3-Dimethylcyclohexanone

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H8(=O)
CAS Number:
Molecular Weight:
126.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

refractive index

n20/D 1.449 (lit.)

bp

174-175 °C (lit.)

density

0.909 g/mL at 25 °C (lit.)

SMILES string

CC1(C)CCCC(=O)C1

InChI

1S/C8H14O/c1-8(2)5-3-4-7(9)6-8/h3-6H2,1-2H3

InChI key

ZVJQBBYAVPAFLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.9 °F - closed cup

Flash Point(C)

59.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service