Skip to Content
Merck
All Photos(1)

Key Documents

390607

Sigma-Aldrich

1-Adamantanecarboxamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO
CAS Number:
Molecular Weight:
179.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

powder

mp

188-190 °C (lit.)

functional group

amide

SMILES string

NC(=O)C12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C11H17NO/c12-10(13)11-4-7-1-8(5-11)3-9(2-7)6-11/h7-9H,1-6H2,(H2,12,13)/t7-,8+,9-,11-

InChI key

CKBZJTAMRPPVSR-KJZNFTALSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V L Andronova
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic], 41(7-8), 26-30 (1996-01-01)
Amide of 1-adamantane carboxylic acid (AACA), the same as remantadine, was shown to have no antiviral action and any influence on the efficiency of the Sindbis virus adsorption. However, it significantly inhibited the virus specific synthesis of RNA and protein.
L Q Gu et al.
Science (New York, N.Y.), 291(5504), 636-640 (2001-02-07)
We describe a heptameric protein pore that has been engineered to accommodate two different cyclodextrin adapters simultaneously within the lumen of a transmembrane beta barrel. The volume between the adapters is a cavity of approximately 4400 cubic angstroms. Analysis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service