Skip to Content
Merck
All Photos(1)

Documents

232173

Sigma-Aldrich

S,S-Diphenylsulfilimine monohydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2S=NH · H2O
CAS Number:
Molecular Weight:
219.30
Beilstein:
2043848
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

67-71 °C (lit.)

SMILES string

[H]O[H].N=S(c1ccccc1)c2ccccc2

InChI

1S/C12H11NS.H2O/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12;/h1-10,13H;1H2

InChI key

YLGYIQQZVOCMDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics and mechanism of reaction of S,S-diphenylsulfilimine with 1-fluoro-2,4-dinitrobenzene, 1-chloro-2,4-dinitrobenzene, 2-chloro-3-nitropyridine, 2-chloro-5-nitropyridine and 2-chloro-3,5-dinitropyridine has been investigated.

Application

S,S-Diphenylsulfilimine monohydrate is a versatile synthetic tool and has been used in preparation of:
  • N-(5-nitrosouracil-6-yl)sulfilimines
  • N-(1,3-dimethyl-6-uracilyl)-S,S-diphenylsulfilimines
  • N-(5-formyl-1,3-dimethyl-6-uracilyl)-S,S-diphenylsulfilimines

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ring closure reactions of ?-nitroso-, ?-acyl-, and ?-thiocarbamoyl-a, ?-unsaturated sulfilimines. Synthesis of [1, 2, 5] oxadiazolo [3, 4-d]-, isoxazolo [3, 4-d]-, and isothiazolo [3, 4-d] pyrimidine derivatives from uracils.
Matsumoto N and Takahashi M.
Tetrahedron, 58(50), 10073-10079 (2002)
Kinetics and mechanism of the reaction of S, S-diphenylsulfilimine with a series of aryl halides.
Sandall JB and Steel ND.
J. Chem. Soc. Perkin Trans. II, 3, 513-516 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service