Skip to Content
Merck
All Photos(1)

Documents

127655

Sigma-Aldrich

Alizarin Complexone dihydrate

indicator grade, Dye content 95 %

Synonym(s):

(3,4-Dihydroxy-2-anthraquinonyl)methyliminodiacetic acid dihydrate, 3,4-Dihydroxyanthraquinon-2-yl-methylimino-diacetic acid, Alizarin Fluorine Blue, Alizarin-3-methylamine-N,N-diacetic acid, Alizarin-3-methyliminodiacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15NO8
CAS Number:
Molecular Weight:
385.32
Beilstein:
2190028
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

indicator grade

composition

Dye content, 95%

mp

190 °C (dec.) (lit.)

λmax

427 nm

Application

For the colorimetric determination of fluoride.

Legal Information

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service