Skip to Content
Merck
All Photos(1)

Key Documents

SRP3012

Sigma-Aldrich

Beta D-1 (47 aa) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

DEFB1, beta-Defensin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

5.0 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

antibiotic activity spectrum

fungi
mycobacteria
viruses

Mode of action

cell membrane | interferes

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DEFB1(1672)

General description

Chemical structure: peptide
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The α-defensins are distinguished from the β-defensins by the pairing of their three disulfide bonds. To date, four human β-defensins have been identified; BD-1, BD-2, BD-3 and BD-4. β-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they are chemoattractant towards immature dendritic cells and memory T cells. The β-defensin proteins are expressed as the C-terminal portion of precursors and are released by proteolytic cleavage of a signal sequence and, in the case of BD-1 (36a.a.), a propeptide region. b-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds. β-defensins are 3-5 kDa peptides ranging in size from 33-47 amino acid residues. Recombinant human BD-1 is a 5.0 kDa protein containing 47 amino acid residues.

Biochem/physiol Actions

Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. Recombinant human BD-1 is a 5.0 kDa protein containing 47 amino acid residues.

Sequence

GNFLTGLGHR SDHYNCVSSG GQCLYSACPI FTKIQGTCYR GKAKCCK

Physical form

Lyophilized from 10 mM Sodium Citrate, pH 4.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in 10 mM Acetic Acid to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert I Lehrer et al.
Current opinion in immunology, 14(1), 96-102 (2002-01-16)
During the past year, novel beta-defensins of mice and men have been identified, together with a novel defensin subfamily (the circular or 'theta' minidefensins) in primates. Insight into the evolution of defensins has been obtained from structural studies, and several
Y Q Tang et al.
Science (New York, N.Y.), 286(5439), 498-502 (1999-10-16)
Analysis of rhesus macaque leukocytes disclosed the presence of an 18-residue macrocyclic, tridisulfide antibiotic peptide in granules of neutrophils and monocytes. The peptide, termed rhesus theta defensin-1 (RTD-1), is microbicidal for bacteria and fungi at low micromolar concentrations. Antibacterial activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service