Skip to Content
Merck
All Photos(1)

Documents

PHR3970

Clonazepam Related Compound A

new

certified reference material, pharmaceutical secondary standard

Synonym(s):

3-Amino-4-(2-chlorophenyl)-6-nitro-2(1H)-quinolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10ClN3O3
CAS Number:
Molecular Weight:
315.71
Beilstein:
5596577
MDL number:

grade

certified reference material
pharmaceutical secondary standard

Agency

traceable to USP 1140327

API family

clonazepam

packaging

pkg of 30MG

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

NC1=C(C2=C(Cl)C=CC=C2)C3=C(NC1=O)C=CC([N+]([O-])=O)=C3

InChI

1S/C15H10ClN3O3/c16-11-4-2-1-3-9(11)13-10-7-8(19(21)22)5-6-12(10)18-15(20)14(13)17/h1-7H,17H2,(H,18,20)

InChI key

GYAWOVGCEQLWHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service