Merck
  • Recent research on 3-phenyllactic acid, a broad-spectrum antimicrobial compound.
Recent research on 3-phenyllactic acid, a broad-spectrum antimicrobial compound.
Applied microbiology and biotechnology (2012-07-12)
Wanmeng Mu, Shuhuai Yu, Lanjun Zhu, Tao Zhang, Bo Jiang
ABSTRACT
3-Phenyllactic acid (PLA), which is an organic acid widely existing in honey and lactic acid bacteria fermented food, can be produced by many microorganisms, especially lactic acid bacteria. It was proved as an ideal antimicrobial compound with broad and effective antimicrobial activity against both bacteria and fungi. In addition, it could be used as feed additives to replace antibiotics in livestock feeds. This article presented a review of recent studies on the existing resource, antimicrobial activity, and measurement of PLA. In addition, microorganism strains and dehydrogenases producing PLA were reviewed in detail, the metabolic pathway and regulation of PLA synthesis in LAB strains were discussed, and high-level bioproduction of PLA by microorganism fermentation was also summarized.
Sigma-Aldrich
D-(+)-3-Phenyllactic acid, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Phenyllactic acid, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-(−)-3-Phenyllactic acid, 98%