MilliporeSigma
  • Antifungal compounds from cultures of dairy propionibacteria type strains.
Antifungal compounds from cultures of dairy propionibacteria type strains.
FEMS microbiology letters (2007-04-26)
Helena Lind, Jörgen Sjögren, Suresh Gohil, Lennart Kenne, Johan Schnürer, Anders Broberg
ABSTRACT
Antifungal compounds from cultures of five type strains of dairy propionibacteria, as well as from the cultivation medium, were studied. Cell-free supernatants and medium were fractionated by C(18) solid phase extraction. The aqueous 95% acetonitrile fractions were analyzed by GC-MS or subjected to reversed-phase HPLC, to identify, quantify or isolate antifungal substances. The resulting HPLC fractions were screened for antifungal activity against the mold Aspergillus fumigatus and the yeast Rhodotorula mucilaginosa. Active fractions were further separated by HPLC and the structures of the compounds were determined by spectroscopic and chromatographic methods. All five strains produced 3-phenyllactic acid, at concentrations ranging from 1.0 microg mL(-1) (Propionibacterium freudenreichii ssp. shermanii) to 15.1 microg mL(-1) (Propionibacterium thoenii), and at L/D -ratios ranging from 2 : 3 (Propionibacterium acidipropionici) to 9 : 1 (Propionibacterium freudenreichii). A number of active compounds found in cultures of propionibacteria were also present in noninoculated growth medium: two antifungal diketopiperazines, cyclo(L-Phe-L-Pro) and cyclo(L-Ile-L-Pro), and seven antifungal linear peptides. Three of the linear peptides corresponded to sequences found in the medium component casein, suggesting their origin from this component, whereas the diketopiperazines were suggested to be formed from medium peptides by heat treatment.
Sigma-Aldrich
D-(+)-3-Phenyllactic acid, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Phenyllactic acid, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-(−)-3-Phenyllactic acid, 98%