Skip to Content
Merck

Z744112

Direct Thermal Cryo-Tags®

white, W × L 1.69 in. × 0.75 in., pack of 500 ea (labels per rolllabels)

Synonym(s):

cryogenic vial labels, freezer labels, Diversified Biotech DTCR-7500, cool storage labels, thermal labels

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
55121611
NACRES:
NB.43
Manufacturer/tradename:
Diversified Biotech DTCR-7500
Material:
white
Packaging:
pack of 500 ea (labels per roll
labels)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

white

packaging

pack of 500 ea (labels per roll
labels)

manufacturer/tradename

Diversified Biotech DTCR-7500

technique(s)

cryopreservation: suitable

W × L

1.69 in. × 0.75 in.

General description

Temperature Range: 70°C to -196°C

Application

Cryogenics

Legal Information

Cryo-Tags is a registered trademark of Diversified Biotech


Still not finding the right product?

Explore all of our products under Direct Thermal Cryo-Tags®




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library