Merck
Z659215
Bandelin SONOPULS ultrasonic homogenizer probes
VS 70 T extended probe for use with HD 2070 homogenizer and SG70G horn or HD 2200 homogenizer and SG 213G horn
Synonym(s):
long probe sonicator, ultrasonic probe, Bandelin 494, Ultrasonic homogenizer tips, Ultrasonic processing probes
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.74
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
titanium (Ti-6AI-4V alloy)
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 494
availability
available only in EU
diam. × L
13 mm × 126 mm
diam.
≥17 mm
volume
10-200 mL , HD 2070, 20-900 mL , HD 2200
General description
Amplitude: HD 2070 = 60 mmss/HD 2200 = 140 mmss
For use with HD 2070 homogenizer and SG70G horn or HD 2200 homogenizer and SG 213G horn. Especially used for sonification of ceramic suspensions or when preparing samples for grain size analysis.
Transmit mechanical longitudinal waves into the sample. They are thermoresistant, can be treated in autoclaves and are resistant to corrosive media. Sample volume, diameter of the processing vessel and the required amplitude determine the selection of the unit and the type of probe. The higher the amplitude, the more intense the sonication.
Using the product image (from left to right):
MS 72 micro tip
MS 73 micro tip
KE 76 tapered tip
VS 70 T extended probe
VS 200 T extended probe
TT 13 flat tip
TT 19 flat tip
TT 25 flat tip
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)