About This Item
NACRES:
NB.12
UNSPSC Code:
41122800
Manufacturer/tradename:
Heathrow Scientific 120817
Feature:
to hold 20 x 10-17 mm tubes OR 12 x 25 - 30 mm tubes
Material:
blue polypropylene (/natural)
Packaging:
pkg of 1 ea
material
blue polypropylene (/natural)
feature
to hold 20 x 10-17 mm tubes OR 12 x 25 - 30 mm tubes
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 120817
L × W × H
309 mm × 107 mm × 77 mm
well diam. × diam. × diam.
10-13 mm × 13-17 m2 × 25-30 mm
General description
The OneRack® Multi is the most flexible tube rack designed for use with a multitude of tube diameters and heights. Each rack comes with 1 base and 6 inserts. Of the 6 inserts, there are two of each size (10-13mm, 13-17mm, 25-30 mm).
Features and Benefits
- Interchangeable tube rack inserts hold multiple tube sizes firmly in place when alternating between sample vessels
- Inserts can be pivoted in either an upper or lower position to securely accommodate the size and height of tube being used
- Open design allows for better filling visibility of tube graduations when filling
- Rack wells have a unique teardrop design that enables better retention of tubes over traditional designs
- Able to rotate the tube rack insert to an angle position to facilitate more comfortable pipetting
Legal Information
OneRack is a registered trademark of Heathrow Scientific LLC
Still not finding the right product?
Explore all of our products under OneRack® Multi
Choose from one of the most recent versions:
Already Own This Product?