Skip to Content

HS120072

TrueNorth® Mini Cooler

for tubes, 0.5 ‑ 2 mL, with gel-filled lid, temp. limit 0 °C, size 32 wells, green polycarbonate

Synonym(s):

cooling tube rack, tube chiller

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41103025
Manufacturer/tradename:
Heathrow Scientific
Feature:
for tubes 0.5 ‑ 2 mL
Material:
green polycarbonate
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

green polycarbonate

feature

for tubes 0.5 ‑ 2 mL

manufacturer/tradename

Heathrow Scientific

parameter

0 °C temp. limit

L × W × H

245 × 105 × 100 mm

size

32 wells

General description

Protect temperature sensitive samples at the bench or in the freezer. Mini Coolers are ideal for keeping PCR reagents, enzymes, and other biochemical reagents cool while working at the bench. Filled with insulating gel, the mini coolers can also be used for ccontrolled sample freezing and for longer temperature retention samples.

Features and Benefits

  • polycarbonate construction with rubber non-slip feet
  • gridded and keyed one-position lids for easy sample identification
  • 0.5 mL tube adapters supplied as standard with the 0.5-2.0 mL tube size versions
  • temperature retention graph supplied with each of the 7 styles
  • versions available or 0.5 and 1.5/2.0 mL microcentrifuge tubes and cyrogenic vials and a 96-place version for 0.2 mL tubes and assay plates

Legal Information

TrueNorth is a registered trademark of Heathrow Scientific LLC


Still not finding the right product?

Explore all of our products under TrueNorth® Mini Cooler




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library