Skip to Content
Merck

HS120036

81-Well microtube storage box, for 1.5-2.0 mL tubes

green, polypropylene

Synonym(s):

microtube storage, vial box

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.12
UNSPSC Code:
41123300
Manufacturer/tradename:
Heathrow Scientific HS120036
Feature:
Holds 81 x 1.5-2.0 mL tubes
Material:
polypropylene
Packaging:
pkg of 5
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene

feature

Holds 81 x 1.5-2.0 mL tubes

packaging

pkg of 5

manufacturer/tradename

Heathrow Scientific HS120036

size

130x130x47 mm

color

green

General description

Compact footprint for microtube and cryogenic vial storage with easy open, friction fit lid. Translucent vibrant colors with a frosted labelling area designed for quick visual identification.

Features and Benefits

  • Compact design conserves valuable bench space, allowing efficient lab organization.
  • Polypropylene construction provides durability and resistance to chemicals, ensuring long-lasting use in the lab environment.
  • Autoclavable material supports easy sterilization, promoting a contamination-free workspace.
  • Color-coded options (assorted colors) enable quick identification and organization of samples, enhancing workflow efficiency.
  • Secure snap closure with a hinged lid ensures safe storage of tubes, minimizing the risk of accidental spills.
  • Stores 81 tubes (1.5-2.0 mL) per box, optimizing sample capacity.Uniform well layout with location identification supports easy tracking of sample placement.
  • Translucent design with a frosted labeling area facilitates labeling and sample visibility.
  • Debossed wave design adds a distinct aesthetic while maintaining functionality.
  • Safe for low-temperature storage, expanding usability for cryogenic samples.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library