Skip to Content
Merck

137267

Hypoxanthine, monosodium salt

EMPROVE® EVOLVE

Synonym(s):

Hypoxanthine monosodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C5H3N4O · Na
Molecular Weight:
158.09
UNSPSC Code:
12352200
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

Quality Level

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin and microbial, tested

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
137267010004065267289441
137267100004065267289458

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service