Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
biological source
human
grade
certified reference material
agency
BCR®
manufacturer/tradename
JRC
application(s)
clinical testing
format
matrix material
storage temp.
−20°C
Other Notes
Sales restrictions may apply
Legal Information
BCR is a registered trademark of European Commission
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.