Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
6D8, monoclonal
Application:
FACS, ICC, IP, WB, inhibition assay
Citations:
-
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
6D8, monoclonal
species reactivity
human
packaging
antibody small pack of 25 μL
technique(s)
flow cytometry: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable, inhibition assay: suitable, western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... DSG2(1829)
General description
Desmoglein-2 (UniProt: Q14126; also known as Cadherin family member 5, HDGC) is encoded by the DSG2 (also known as CDHF5) gene (Gene ID: 1829) in human. Desmogleins are protein substances located in and on the surface of keratinocytes. They are present in specialized cell-cell adhesion structures enriched in tissues that undergo substantial mechanical stress. These proteins have been shown to be a critical factor in cell-to-cell adhesion. Desmoglein-2 is a single-pass type I membrane protein that is found in all tissues and in carcinomas. It serves as a component of intercellular desmosome junctions. It is involved in the interaction of plaque proteins and intermediate filaments mediating cell-cell adhesion. It is synthesized with a signal peptide (aa 1-23) and a propeptide (aa 24-49) that are cleaved off in the mature form. It contains an extracellular domain (aa 50-609), a transmembrane domain (aa 610-634), and a cytoplasmic domain (aa 635-1118). Mutations in DSG2 gene have been linked to familial type of arrhythmogenic right ventricular dysplalsia that is characterized y infiltration of adipose and fibrous tissue into the right ventricle and loss of myocardial cells, resulting in ventricular and supraventricular arrhythmias.
~165 kDa observed; 122.29 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Immunogen
Supernatant from cell membranes extract from A-431 cells.
Application
Anti-Desmoglein 2, clone 6D8, Cat. No. MABT1507, is a mouse monoclonal antibody that detects Desmoglein-2 and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibition assays, and Western Blotting,
Flow Cytometry Analysis: A representative lot detected Desmoglein 2 in Flow Cytometry applications (Tan, L.Y., et. al. (2016). Oncotarget. 7(29):46492-46508).
Western Blotting Analysis: A 1:125 dilution from a repesentative lot detected Desmoglein 2 in HaCaT cell lysates.
Flow Cytometry Analysis: 1 µg from a representative lot detected Desmoglein2 in SK-MEL-28.
Western Blotting Analysis: A representative lot detected Desmoglein 2 in Western Blotting applications (Wahl, J.K. 3rd, et. al. (2002). Hybrid Hybridomics. 21(1):37-44).
Immunocytochemistry Analysis: A representative lot detected Desmoglein 2 in Immunocytochemistry applications (Wahl, J.K. 3rd, et. al. (2002). Hybrid Hybridomics. 21(1):37-44; Tan, L.Y., et. al. (2016). Oncotarget. 7(29):46492-46508).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Desmoglein 2 in Immunoprecipitation applications (Wahl, J.K. 3rd, et. al. (2002). Hybrid Hybridomics. 21(1):37-44).
Inhibition Analysis: A representative lotinhibited the attachment of adenovirus (Ad) serotypes Ad3.(Wang, H., et. al. (2011). Nat Med. 17(1):96-104).
Western Blotting Analysis: A 1:125 dilution from a repesentative lot detected Desmoglein 2 in HaCaT cell lysates.
Flow Cytometry Analysis: 1 µg from a representative lot detected Desmoglein2 in SK-MEL-28.
Western Blotting Analysis: A representative lot detected Desmoglein 2 in Western Blotting applications (Wahl, J.K. 3rd, et. al. (2002). Hybrid Hybridomics. 21(1):37-44).
Immunocytochemistry Analysis: A representative lot detected Desmoglein 2 in Immunocytochemistry applications (Wahl, J.K. 3rd, et. al. (2002). Hybrid Hybridomics. 21(1):37-44; Tan, L.Y., et. al. (2016). Oncotarget. 7(29):46492-46508).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Desmoglein 2 in Immunoprecipitation applications (Wahl, J.K. 3rd, et. al. (2002). Hybrid Hybridomics. 21(1):37-44).
Inhibition Analysis: A representative lotinhibited the attachment of adenovirus (Ad) serotypes Ad3.(Wang, H., et. al. (2011). Nat Med. 17(1):96-104).
Research Category
Cell Structure
Cell Structure
Biochem/physiol Actions
Clone 6D8 specifically detects Desmoglein-2 in human cells.
Physical form
Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.
Preparation Note
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
Evaluated by Immunocytochemistry in A431 cell line.
Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected Desmoglein 2 in A431 cell line.
Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected Desmoglein 2 in A431 cell line.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.