GF35446316
Titanium
rod, 100mm, diameter 20mm, annealed, 99.6+%
Synonym(s):
Titanium, TI007940
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99.6%
form
rod
autoignition temp.
860 °F
manufacturer/tradename
Goodfellow 354-463-16
resistivity
42.0 μΩ-cm, 20°C
L × diam.
100 mm × 20 mm
bp
3287 °C (lit.)
mp
1660 °C (lit.)
density
4.5 g/mL at 25 °C (lit.)
SMILES string
[Ti]
InChI
1S/Ti
InChI key
RTAQQCXQSZGOHL-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Aaron F Cipriano et al.
Journal of biomedical nanotechnology, 10(4), 660-668 (2014-04-17)
The objective of this study was to investigate the adhesion and morphology of bone marrow derived stromal cells (BMSCs) on bulk titanium (Ti) substrates with precisely-patterned surfaces consisting of groove-based gratings with groove widths ranging from 50 micro m down
Shinya Kato et al.
Journal of nanoscience and nanotechnology, 14(5), 3285-3291 (2014-04-17)
Microcorpuscular titanium dioxide (TiO2), a useful sunscreen agent, photocatalyzes generation of reactive oxygen species (ROS). We assessed protective effects of fullerene-C60 derivatives or microcolloidal platinum (Pt) against ultraviolet ray (UV)-irradiation in the presence of TiO2 in vitro. UV-irradiation (8 J/cm2
Evsen Tamam et al.
The Journal of oral implantology, 40(2), 153-159 (2014-05-02)
Implant failure is more likely to occur in persons with medically compromising systemic conditions, such as diabetes related to high blood glucose levels and inflammatory diseases related to pH levels lower than those in healthy people. The aim of this
Wei-Qiang Yu et al.
Journal of nanoscience and nanotechnology, 14(6), 4387-4393 (2014-04-18)
The TiO2 nanotubes by anodization have been extensively studied for medical implant and orthopedic applications because of enhancing bone development. In the present study, a new nano-foveolae structure verified by SEM and AFM was prepared by simulating the nanotubes exfoliation
Shaoming Yang et al.
Journal of nanoscience and nanotechnology, 14(6), 4292-4296 (2014-04-18)
The structural and functional characterizations of titanium nitride (TiN) advanced ceramic microsprings (CMSs), with a coil diameter of several micrometers and synthesized by chemical vapor deposition (CVD) were investigated by microscopy techniques. The CMSs were sufficiently mechanically elastic for extension
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service