Skip to Content
Merck

489239

Ethane-d1

98 atom % D

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
CH3CH2D
CAS Number:
Molecular Weight:
31.08
UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:
Isotopic purity:
98 atom % D
Mass shift:
M+1


vapor density

1.05 (vs air)

Quality Segment

vapor pressure

37.95 atm ( 21.1 °C)

isotopic purity

98 atom % D

autoignition temp.

881 °F

bp

−88 °C (lit.)

mp

−172 °C (lit.)

mass shift

M+1

SMILES string

[2H]CC

InChI

1S/C2H6/c1-2/h1-2H3/i1D

InChI key

OTMSDBZUPAUEDD-MICDWDOJSA-N

Other Notes

Stainless steel control valves Z146951, Z187224 or either of stainless steel regulators Z512966 and Z512974 is recommended.


pictograms

FlameGas cylinder

signalword

Danger

hcodes

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class

2A - Gases

wgk

WGK 3

flash_point_f

-211.0 °F - closed cup

flash_point_c

-135.00 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library