MTCA9004WR
MTC™ Bio Autoclave Bag Wire Rack Stand
holds autoclave bags 8.5in. x 11in. (21.6 x 27.9cm)
Synonym(s):
biohazard bag holder, biohazard bag lab stand
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
steel
feature
holds autoclave bags 8.5in. x 11in. (21.6 x 27.9cm)
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio A9004-WR
General description
Wire rack stand for 8.5 x 11" autoclave bags
Features and Benefits
- sturdy steel construction
- autoclavable
Legal Information
MTC is a trademark of MTC Bio
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service