Skip to Content
Merck
All Photos(1)

Key Documents

EHU096471

Sigma-Aldrich

MISSION® esiRNA

targeting human DCN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAGCCTCATTTGAATGTGTGAATTCAATACAGGCTATGTAAAATTTTTACTAATGTCATTATTTTGAAAAAATAAATTTAAAAATACATTCAAAATTACTATTGTATACAAGCTTAATTGTTAATATTCCCTAAACACAATTTTATGAAGGGAGAAGACATTGGTTTGTTGACAATAACAGTACATCTTTTCAAGTTCTCAGCTATTTCTTCTACCTCTCCCTATCTTACATTTGAGTATGGTAACTTATGTCATCTATGTTGAATGTAAGCTTATAAAGCACAAAGCATACATTTCCTGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Jiang et al.
International immunopharmacology, 69, 373-381 (2019-02-19)
Kaempferol is a kind of bioflavonoid exerts diverse pharmacological activities, including anti-apoptotic and anti-inflammatory activities. Kaempferol has been recognized as an effective agent for alleviating the clinical symptoms of osteoarthritis (OA). This study aimed to provide evidence that Kaempferol has
Yoshihiro Joshua Ono et al.
The Journal of endocrinology, 223(2), 203-216 (2014-09-24)
Dienogest, a synthetic progestin, has been shown to be effective against endometriosis, although it is still unclear as to how it affects the ectopic endometrial cells. Decorin has been shown to be a powerful endogenous tumor repressor acting in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service