CLS3231
Corning® CellCube®
10 layer, TCT, with 6” Silicone tubing and Aseptic Connectors, sterile, bag of 1 ea, case of 2 ea
Synonym(s):
cell culture vessel
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polystyrene
silicon tubing (6′′)
sterility
sterile
packaging
bag of 1 ea
case of 2 ea
manufacturer/tradename
Corning 3231
surface area
8500 cm2 , cell growth area
tubing O.D.
6 in.
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Introducing Corning′s Closed System Cell Cube – a new closed system offering designed to help reduce the risk of adventitious contamination. Our new offering of CellCube 10, 25, 100 modules, circulation loops, and connectors allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik® and MPC connectors.
Application
Adherent Cell Scale Up
Features and Benefits
- 8,500 cm2 Surface Area
- Corning TCT surface treatment
- Sterility Assurance Level(SAL) of 10-6
- AseptiQuik® G Connector
Legal Information
AseptiQuik is a registered trademark of Colder Products Company
CELLCUBE is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service