Skip to Content
Merck
All Photos(1)

Key Documents

N1-S1

90011902, rat liver, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

N1-S1, 90011902

biological source

rat liver

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat hepatoma

Cell Line Description

Established from the ascitic fluid of a Novikoff hepatoma. The cells can be synchronised for cell cycle studies. N1-S1 has also been used for studies of pyrimidine metabolism, DNA synthesis and methylation and ribosomal RNA maturation.

Application

Cell cycle, metabolic and mutation studies

Culture Medium

Swimm′s S-77 + 2mM Glutamine + 0.1% Pluronic F68 +10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 4x10,000 to 1,000,000 cells/ml; 5% CO2; 37°C. Subculture every 3-4 days by centrifugation or medium addition.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service