Skip to Content
Merck
All Photos(1)

Key Documents

AV38863

Sigma-Aldrich

Anti-LZTR1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Leucine-zipper-like transcription Regulator 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

95 kDa

species reactivity

bovine, rabbit, human, horse, guinea pig, dog, mouse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LZTR1(8216)

Immunogen

Synthetic peptide directed towards the C terminal region of human LZTR1

Biochem/physiol Actions

LZTR1 is a transcription regulator and a member of BTB-kelch superfamily of proteins that regulate cellular processes such as migration, gene expression and cell morphology. It is localized to Golgi complex and has been reported as Golgi matrix-associated protein. Deletions in LZTR1 gene has been observed in DiGeorge syndrome patients.

Sequence

Synthetic peptide located within the following region: GFYNNRLQAYCKQNLEMNVTVQNVLQILEAADKTQALDMKRHCLHIIVHQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanju G Nacak et al.
The Journal of biological chemistry, 281(8), 5065-5071 (2005-12-17)
Members of the BTB-kelch superfamily play important roles during fundamental cellular processes, such as the regulation of cell morphology, migration, and gene expression. The BTB-kelch protein LZTR-1 is deleted in the majority of DiGeorge syndrome patients and is believed to
H Kurahashi et al.
Human molecular genetics, 4(4), 541-549 (1995-04-01)
The region commonly deleted in DiGeorge syndrome (DGS) has been localized at 22q11.1-q11.2 with the aid of a high resolution banding technique. A 22q11 specific plasmid library was constructed with a microdissection and microcloning method. Dosage analysis proved three of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service