Merck
  • Electrochemical and electron microscopic characterization of Super-P based cathodes for Li-O2 batteries.
Electrochemical and electron microscopic characterization of Super-P based cathodes for Li-O2 batteries.
Beilstein journal of nanotechnology (2013-11-10)
Mario Marinaro, Santhana K Eswara Moorthy, Jörg Bernhard, Ludwig Jörissen, Margret Wohlfahrt-Mehrens, Ute Kaiser
ABSTRACT
Aprotic rechargeable Li-O2 batteries are currently receiving considerable interest because they can possibly offer significantly higher energy densities than conventional Li-ion batteries. The electrochemical behavior of Li-O2 batteries containing bis(trifluoromethane)sulfonimide lithium salt (LiTFSI)/tetraglyme electrolyte were investigated by galvanostatic cycling and electrochemical impedance spectroscopy measurements. Ex-situ X-ray diffraction and scanning electron microscopy were used to evaluate the formation/dissolution of Li2O2 particles at the cathode side during the operation of Li-O2 cells.
Sigma-Aldrich
Bis(trifluoromethane)sulfonimide lithium salt, 99.95% trace metals basis