MilliporeSigma
  • ORP5 localizes to ER-lipid droplet contacts and regulates the level of PI(4)P on lipid droplets.
ORP5 localizes to ER-lipid droplet contacts and regulates the level of PI(4)P on lipid droplets.
The Journal of cell biology (2019-10-28)
Ximing Du, Linkang Zhou, Yvette Celine Aw, Hoi Yin Mak, Yanqing Xu, James Rae, Wenmin Wang, Armella Zadoorian, Sarah E Hancock, Brenna Osborne, Xiang Chen, Jia-Wei Wu, Nigel Turner, Robert G Parton, Peng Li, Hongyuan Yang
ABSTRACT
Lipid droplets (LDs) are evolutionarily conserved organelles that play important roles in cellular metabolism. Each LD is enclosed by a monolayer of phospholipids, distinct from bilayer membranes. During LD biogenesis and growth, this monolayer of lipids expands by acquiring phospholipids from the endoplasmic reticulum (ER) through nonvesicular mechanisms. Here, in a mini-screen, we find that ORP5, an integral membrane protein of the ER, can localize to ER-LD contact sites upon oleate loading. ORP5 interacts with LDs through its ligand-binding domain, and ORP5 deficiency enhances neutral lipid synthesis and increases the size of LDs. Importantly, there is significantly more phosphatidylinositol-4-phosphate (PI(4)P) and less phosphatidylserine (PS) on LDs in ORP5-deficient cells than in normal cells. The increased presence of PI(4)P on LDs in ORP5-deficient cells requires phosphatidylinositol 4-kinase 2-α. Our results thus demonstrate the existence of PI(4)P on LDs and suggest that LD-associated PI(4)P may be primarily used by ORP5 to deliver PS to LDs.
Sigma-Aldrich
Anti-OSBPL5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody