Z743536
Richter Anti-Vibration pads
L × W 8.5 in. × 11.0 in., pkg of 2
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
Recommended Products
packaging
pkg of 2
manufacturer/tradename
Diversified Biotech VIBE-2000
L × W
8.5 in. × 11.0 in.
General description
Inexpensive solution which dampens vibrations generated from lab equipment
Creates a stable surface to keep equipment in place
Fits vortexes, shakers, rockers and microscopes
Small, lightweight design
Stack pads for heavier units or position side-by-side forlarger equipment
Creates a stable surface to keep equipment in place
Fits vortexes, shakers, rockers and microscopes
Small, lightweight design
Stack pads for heavier units or position side-by-side forlarger equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)