EP0030128966
Eppendorf® Twin tec PCR plates, 384 wells
blue polypropylene plate, conical bottom colorless wells, pkg of 300 ea
All Photos(1)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
blue polypropylene plate
conical bottom colorless wells
conical bottom colorless wells
size:
384 wells
sterility:
non-sterile
binding type:
low binding surface
feature:
skirt
Recommended Products
material
blue polypropylene plate
conical bottom colorless wells
sterility
non-sterile
feature
skirt
packaging
pkg of 300 ea
manufacturer/tradename
Eppendorf® 0030128966
technique(s)
PCR: suitable
size
384 wells
well maximum volume
45 μm
working volume
-40 μL
binding type
low binding surface
General description
“Eppendorf” Twin tec PCR plates with 384 wells are large scale PCR plates used in high throughput PCR workflows. They have rigid polycarbonate frames, polypropylene wells and raised well rims, ensuring a sturdy plate with effective sealing.
Application
Eppendorf “twin.tec” PCR Plates may be used in a wide variety of applications such as:
- Standard PCR
- Real time PCR
- High throughput PCR
Features and Benefits
- “Eppendorf” Twin tec PCR plates have polypropylene wells with a maximum volume of 40?L.
- Enhanced rim design for efficient barcode scanning and automated workflows.
- The plates can be stacked together enabling convenient storage and handling.
- These Twin tec PCR plates are compatible with most 384- well thermocycler.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)