8.20647
4-Hexylresorcinol
for synthesis
Synonym(s):
4-Hexylresorcinol, 1,3-Dihydroxy-4-hexylbenzene, 1,3-Dihydroxy-4-hexylbenzene
About This Item
Recommended Products
Quality Level
form
powder
potency
550 mg/kg LD50, oral (Rat)
pH
7.7 (20 °C, 0.5 g/L in H2O)
bp
333-335 °C/1013 hPa
mp
64-67 °C
transition temp
flash point >100 °C
bulk density
340 kg/m3
storage temp.
2-30°C
SMILES string
Oc1c(ccc(c1)O)CCCCCC
InChI
1S/C12H18O2/c1-2-3-4-5-6-10-7-8-11(13)9-12(10)14/h7-9,13-14H,2-6H2,1H3
InChI key
WFJIVOKAWHGMBH-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
388.4 °F
Flash Point(C)
198 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service