Merck
  • Study on adsorption of glyphosate (N-phosphonomethyl glycine) pesticide on MgAl-layered double hydroxides in aqueous solution.
Study on adsorption of glyphosate (N-phosphonomethyl glycine) pesticide on MgAl-layered double hydroxides in aqueous solution.
Journal of hazardous materials (2005-07-12)
Feng Li, Yunfeng Wang, Qiaozhen Yang, David G Evans, Claude Forano, Xue Duan
ABSTRACT
MgAl-layered double hydroxides with different interlayer anions (nitrate, carbonate and chloride) were evaluated for their abilities to adsorb the organic pesticide glyphosate (N-phosphonomethyl glycine, Gly). The adsorption isotherms of Gly on layered double hydroxides (LDHs) nitrate were described by the Langmuir equation at lower equilibrium concentration of Gly (C(e)<1.0 mmol/L), and the Gly adsorption capacity on LDHs increased with the layer charge density, i.e. the structural Al3+/Mg2+ ratio. Gly adsorption on LDHs nitrate generally occurred through two processes, external surface adsorption and interlayer anion exchange. The adsorption amount on LDHs at C(e)=1.0 mmol/L decreased in the order of interlayer anions: Cl(-)>NO3(-)>CO3(2-).
Sigma-Aldrich
Magnesium aluminate, single crystal substrate, <111>, 99.99% trace metals basis
Sigma-Aldrich
Magnesium aluminate, single crystal substrate, <110>, 99.99% trace metals basis
Sigma-Aldrich
Magnesium aluminate, spinel, nanopowder, <50 nm particle size (BET)