Skip to Content
Merck
All Photos(1)

Key Documents

634840

Sigma-Aldrich

Magnesium aluminate

single crystal substrate, <110>, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO·Al2O3
CAS Number:
Molecular Weight:
142.27
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.99% trace metals basis

hardness

8.0 mohs

size

10 mm × 10 mm × 0.5 mm

mp

2130 °C (lit.)

density

3.64 g/mL at 25 °C (lit.)

semiconductor properties

<110>

SMILES string

[Mg++].[O-][Al]=O.[O-][Al]=O

InChI

1S/2Al.Mg.4O/q;;+2;;;2*-1

InChI key

VPBIQXABTCDMAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Inert matrix for the transmutation of minor actinides such as americium.[1]

Features and Benefits

Transparent substrate used for bulk microwave and acoustic wave devices. Good substrate for III-V nitride materials.

Physical properties

propagation loss: 6.5 dB/μs; phase velocity: 6500 m/x at (100> shear wave); thermal expansion: 7.45 × 10-6 /°C

Physical form

cubic (a = 8.083 Å)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satit Puttipipatkhachorn et al.
International journal of pharmaceutics, 293(1-2), 51-62 (2005-03-22)
Diclofenac calcium-alginate (DCA) beads were reinforced with different amounts of sodium starch glycolate (SSG) or magnesium aluminum silicate (MAS) and were prepared using ionotropic gelation method. Complex formation of sodium alginate (SA) and SSG or MAS in calcium-alginate beads was
J C Keller et al.
Dental materials : official publication of the Academy of Dental Materials, 13(1), 62-68 (1997-01-01)
The objective of this work was to examine osteoblast-like cell attachment and morphology in vitro to osteoceramic materials with three different surface morphologies. Osteoceramic composite disks were fabricated from tricalcium phosphate and magnesium-aluminate spinel (MgAl2O4) in a 50 vol% ratio.
Tomohito Kameda et al.
Chemosphere, 73(5), 844-847 (2008-07-25)
Magnesium-aluminum oxide (Mg-Al oxide) obtained by thermal decomposition of Mg-Al layered double hydroxide (Mg-Al LDH) effectively removed HCl from gaseous streams. HCl removal was greater in the presence of added water vapor at all temperatures examined and increased with decreasing
Klaassen, F.C.
J. Nuc. Mat., 319, 108-108 (2003)
Tomohito Kameda et al.
Chemosphere, 82(4), 587-591 (2010-11-30)
Although effective treatment of acid gases such as HCl, SO(x), and NO(x) is essential for preventing air pollution, current methods pose other environmental problems such as CaCl₂ leaching, reduced landfill lifetimes, and solid waste production. Here we show that acid

Articles

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service