Merck
  • Thermal activation of non-radiative Auger recombination in charged colloidal nanocrystals.
Thermal activation of non-radiative Auger recombination in charged colloidal nanocrystals.
Nature nanotechnology (2013-02-12)
C Javaux, B Mahler, B Dubertret, A Shabaev, A V Rodina, Al L Efros, D R Yakovlev, F Liu, M Bayer, G Camps, L Biadala, S Buil, X Quelin, J-P Hermier
ABSTRACT
Applications of semiconductor nanocrystals such as biomarkers and light-emitting optoelectronic devices require that their fluorescence quantum yield be close to 100%. However, such quantum yields have not been obtained yet, in part, because non-radiative Auger recombination in charged nanocrystals could not be suppressed completely. Here, we synthesize colloidal core/thick-shell CdSe/CdS nanocrystals with 100% quantum yield and completely quenched Auger processes at low temperatures, although the nanocrystals are negatively photocharged. Single particle and ensemble spectroscopy in the temperature range 30-300 K shows that the non-radiative Auger recombination is thermally activated around 200 K. Experimental results are well described by a model suggesting a temperature-dependent delocalization of one of the trion electrons from the CdSe core and enhanced Auger recombination at the abrupt CdS outer surface. These results point to a route for the design of core/shell structures with 100% quantum yield at room temperature.
Sigma-Aldrich
Lumidot CdS, core-type quantum dots, 5 mg/mL in toluene
Sigma-Aldrich
Cadmium sulfide, powder, 99.995% trace metals basis