Merck
  • Photo-induced self-cleaning and sterilizing activity of Sm3+ doped ZnO nanomaterials.
Photo-induced self-cleaning and sterilizing activity of Sm3+ doped ZnO nanomaterials.
Chemosphere (2012-11-06)
M Saif, H Hafez, A I Nabeel
ABSTRACT
Highly active samarium doped zinc oxide self-cleaning and biocidal surfaces (x mol% Sm(3+)/ZnO where x=0, 1, 2 and 4 mol%) with crystalline porous structures were synthesized by hydrothermal method. Sm(3+)/ZnO thin films were characterized by X-ray diffraction (XRD), transmission electron microscope (TEM), scanning electron microscope (SEM), energy dispersive spectroscopic (EDS), UV-visible diffuse reflectance and fluorescence (FL) spectroscopy. The combination between doping and hydrothermal treatments significantly altered the morphology of ZnO into rod and plate-like nanoshapes structure and enhanced its absorption and emission of ultraviolet radiation. The photo-activity in term of quantitative determination of the active oxidative species (()OH) produced on the thin film surfaces was evaluated using fluorescent probe method. The results showed that, the hydrothermally treated 2.0 mol% Sm(3+)/ZnO film (S2) is the highly active one. The optical, structural, morphology and photo-activity properties of the highly active thin film (S2) make it promising surface for self-cleaning and sterilizing applications.
Sigma-Aldrich
Samarium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Samarium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Samarium, −40 mesh, 99% trace rare earth metals basis