263184
Samarium
ingot, 99.9% trace rare earth metals basis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Sm
CAS Number:
Molecular Weight:
150.36
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99.9% trace rare earth metals basis
form
ingot
reaction suitability
reagent type: catalyst
core: samarium
resistivity
91.4 μΩ-cm, 0°C
bp
1794 °C (lit.)
mp
1074 °C (lit.)
density
7.47 g/mL at 25 °C (lit.)
SMILES string
[Sm]
InChI
1S/Sm
InChI key
KZUNJOHGWZRPMI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Water-react 3
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yi Lu et al.
Optics express, 21(2), 1997-2002 (2013-02-08)
We demonstrate an all-fiber passively Q-switched Yb-doped laser using a piece of Sm-doped fiber as a saturable absorber. The laser was pumped by two 25W, 975 nm fiber coupled diodes and Q-switching was initiated when the ASE generated in the
M Majumder et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(2), 025701-025701 (2012-12-05)
We report resistivity, magnetization and (19)F NMR results in a polycrystalline sample of SmFeAsO(0.86)F(0.14). The resistivity and magnetization data show a sharp drop at 48 K indicating a superconducting transition. The nuclear spin-lattice rate (1/T(1)) and spin-spin relaxation rate (1/T(2))
I Pal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 101, 74-81 (2012-10-27)
The glasses with compositions 20ZnO·(79.5-x)Bi2O3·xSiO2·0.5Sm2O3 (10≤x≤50, mol%) have been synthesized using normal melt-quench technique. Optical absorption and fluorescence spectra of the glasses were recorded at ambient temperature. Judd-Ofelt (J-O) theory has been successfully applied to characterize the absorption and luminescence
Hua Yang et al.
Inorganic chemistry, 52(3), 1275-1284 (2013-01-25)
Sm@C(2v)(3)-C(80) has been separated from the carbon soot produced by electrical arc vaporization of graphite rods doped with Sm(2)O(3) and purified. Its structure has been determined by single crystal X-ray diffraction using cocrystals obtained from either Ni(II)(octaethylporphyrin) (Ni(II)(OEP)) to form
Yongchao Jia et al.
Chemical communications (Cambridge, England), 49(26), 2664-2666 (2013-02-26)
The Eu(2+) → Tb(3+) → Sm(3+) energy transfer scheme has been proposed to realize the sensitization of Sm(3+) ion emission by Eu(2+) ions. Following this energy transfer model, near-UV convertible Sm(3+)-activated orange phosphors have been obtained in Sr3Y(PO4)3:Eu(2+), Tb(3+), Sm(3+)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service