Merck
  • Amino acid sequence of a hypertrehalosaemic neuropeptide from the corpus cardiacum of the cockroach, Nauphoeta cinerea.
Amino acid sequence of a hypertrehalosaemic neuropeptide from the corpus cardiacum of the cockroach, Nauphoeta cinerea.
Biochemical and biophysical research communications (1986-12-15)
G Gäde, K L Rinehart
ABSTRACT
The hypertrehalosaemic factor from the cockroach Nauphoeta cinerea, a decapeptide, has been assigned the structure >Glu-Val-Asn-Phe-Ser-Pro-Gly-Trp-Gly-Thr-NH2. The structure was assigned from its high-resolution fast atom bombardment mass spectrum and metastable studies on the M + H ion and confirmed by solid phase synthesis.
Sigma-Aldrich
pGlu-Val-Asn-Phe-Ser-Pro-Gly-Trp-Gly-Thr-NH2, ≥95% (HPLC)