
Sodium 1-octanesulfonate solution

Synonym(s):
1-Octanesulfonic acid sodium salt
Linear Formula:
CH3(CH2)6CH2SO3Na
CAS Number:
Molecular Weight:
216.27
Products
Brand
Formula Weight
Physical Form
Quality Segment
Package Size
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing