Skip to Content
Merck
N-Phthaloyl-L-glutamic acid 99% (HPLC)

N-Phthaloyl-L-glutamic acid

N-Phthaloyl-L-glutamic acid 99% (HPLC)
Synonym(s):
PhGA
Empirical Formula (Hill Notation):
C13H11NO6
CAS Number:
Molecular Weight:
277.23

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing