Aluminum standard for AAS
Synonym(s):
Aluminum chloride solution
Empirical Formula (Hill Notation):
AlCl3
CAS Number:
Molecular Weight:
133.34
Products
Brand
Formula Weight
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing