(1S,2S)-(2-Amino-1-phenylpropyl)diphenylphosphine
Synonym(s):
(1S,2S)-(2-Diphenylphosphino)-1-methyl-2-phenylethylamine
Empirical Formula (Hill Notation):
C21H22NP
CAS Number:
Molecular Weight:
319.38
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing