Skip to Content
Merck
1-(2-Pyrimidyl)piperazine 98%

1-(2-Pyrimidyl)piperazine

1-(2-Pyrimidyl)piperazine 98%
Synonym(s):
2-(1-Piperazinyl)pyrimidine
Empirical Formula (Hill Notation):
C8H12N4
Molecular Weight:
164.21
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing