Skip to Content
Merck
All Photos(3)

Key Documents

V900474

Sigma-Aldrich

Guanidine thiocyanate

Vetec, reagent grade, ≥97%

Synonym(s):

Guanidinium rhodanide, Guanidinium thiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NH2 · HSCN
CAS Number:
Molecular Weight:
118.16
Beilstein:
3563461
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

≥97%

form

powder or crystals

mp

120-122 °C (lit.)

SMILES string

SC#N.NC(N)=N

InChI

1S/CH5N3.CHNS/c2-1(3)4;2-1-3/h(H5,2,3,4);3H

InChI key

ZJYYHGLJYGJLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chaotropic agent and strong denaturant; solubilizes cells.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kai Y Mak et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 23(9), 1434-1443 (2015-05-23)
Angiotensin converting enzyme 2 (ACE2) which breaks down profibrotic peptide angiotensin II to antifibrotic peptide angiotensin-(1-7) is a potential therapeutic target in liver fibrosis. We therefore investigated the long-term therapeutic effect of recombinant ACE2 using a liver-specific adeno-associated viral genome
Taku Takeda et al.
Bioscience, biotechnology, and biochemistry, 73(2), 366-371 (2009-02-10)
The stability of the oxidized and reduced forms of three homologous cytochromes c from two thermophiles and one mesophile was systematically monitored by means of Soret absorption measurements in the presence of various concentrations of a denaturant, guanidine thiocyanate, at
Yuji Suzuki et al.
Bioscience, biotechnology, and biochemistry, 72(7), 1951-1953 (2008-07-08)
High-quality total RNA was extracted using a cethyltrimethylammonium bromide-containing buffer followed by an acid guanidium thiocyanate-phenol-chloroform treatment from recalcitrant plant tissues such as tree leaves (pine, Norway spruce, ginkgo, Japanese cedar, rose), flowers (rose, Lotus japonicus) and storage tissues (seeds
Johannes Hunger et al.
The journal of physical chemistry. B, 117(2), 615-622 (2012-12-18)
We study the conductance of dilute aqueous solutions for a series of guandinium salts at 298.15 K. The experimental molar conductivities were analyzed within the framework of the Quint-Viallard theory in combination with Debye-Hückel activity coefficients. From this analysis, we
Olesya V Stepanenko et al.
PloS one, 7(11), e48809-e48809 (2012-11-13)
Having a high folding efficiency and a low tendency to aggregate, the superfolder GFP (sfGFP) offers a unique opportunity to study the folding of proteins that have a β-barrel topology. Here, we studied the unfolding-refolding of sfGFP that was induced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service