Skip to Content
Merck
All Photos(1)

Key Documents

V000196

Sigma-Aldrich

Cadmium sulfate 8/3-hydrate

ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSO4 · 8/3H2O
CAS Number:
Molecular Weight:
256.57
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

ACS reagent

SMILES string

[Cd++].[Cd++].[Cd++].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/3Cd.3H2O4S.8H2O/c;;;3*1-5(2,3)4;;;;;;;;/h;;;3*(H2,1,2,3,4);8*1H2/q3*+2;;;;;;;;;;;/p-6

InChI key

AGHQMAQBLOTWPQ-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Cadmium sulfate 8/3-hydrate can be used:
  • To prepare a standard solution of cadmium, used in the study of the application of amorphous calcium phosphate composite in cadmium adsorption.
  • As a precursor reagent in the preparation of mesoporous CdSe photocatalyst, used in CO2 conversion.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Target Organs

Bone,Kidney,Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photocatalytic CO2 conversion on highly ordered mesoporous materials: Comparisons of metal oxides and compound semiconductors
Lee Y, et al.
Applied Catalysis. B, Environmental, 224, 594-601 (2018)
One-pot synthesis of amorphous calcium phosphate/Fe3O4 composites and the application in the removal of cadmium
Trang T, et al.
Journal of Environmental Chemical Engineering, 103653-103653 (2020)
Catriona O'Sullivan et al.
Chemical communications (Cambridge, England), 47(9), 2655-2657 (2011-01-12)
A multi component assembly consisting of the redox protein cytochrome c (cyt c) immobilised onto vertically aligned gold tipped semiconductor nanorods is described. Cyt c was successfully immobilised using a thiol linker. A faradaic response demonstrated that the protein is
P Voua Otomo et al.
Ecotoxicology (London, England), 19(2), 362-368 (2009-09-23)
Since life-cycle studies showed that the earthworm species Eisenia fetida can develop increased tolerance after long-term exposure to a sub-lethal concentration of Cd in the laboratory, we assessed both the cytotoxicity and genotoxicity of Cd in a long-term Cd-exposed population.
Laura Saa et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(21), 6187-6192 (2010-05-01)
We have developed an analytical assay to detect the enzymatic activity of acetylcholine esterase and alkaline phosphatase based on the generation of quantum dots by enzymatic products. Acetylcholine esterase converts acetylthiocholine into thiocholine. The latter enhances the rate of decomposition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service