Skip to Content
Merck
All Photos(1)

Key Documents

58378

Supelco

SUPELCOSIL LC-PCN (5 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Synonym(s):

MTO-SUPELCOSIL LC-PCN 5UM 25CMX4.6MM HPLC COLUMN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

SUPELCOSIL LC-PCN HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

Agency

suitable for USP L10

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

4% carbon loading

parameter

0-70 °C temperature
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 3.5 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

cyano phase

particle size

5 μm

pore size

120 Å pore size

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

The LC-PCN cyanopropyl bonded phase columns are preconditioned for fast and reliable analyses of tricyclic antidepressants.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W N Lin
Therapeutic drug monitoring, 9(3), 337-342 (1987-09-01)
A simple, specific, and sensitive high-performance liquid chromatography (HPLC) method has been developed for the routine monitoring in serum of the benzodiazepine anticonvulsant, clonazepam. Serum spiked with internal standard, methylclonazepam, was vortex-mixed for 1 min with chloroform at an alkaline
W N Lin et al.
Therapeutic drug monitoring, 9(4), 448-455 (1987-12-01)
An analytical procedure has been developed for the simultaneous separation and quantitation of amitriptyline (AMI), imipramine (IMI), doxepin (DOX), trimipramine (TRI), desipramine (DES), nortriptyline (NOR), desmethyldoxepin (DMD), and protriptyline (PRO) in serum using N-propionylprocainamide (NPPA) as an internal standard. Serum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service