Skip to Content
Merck
All Photos(1)

Documents

508500

Supelco

Crimp cap, 20 mm, aluminum

open center, silver aluminum, Chromacol 20-ACB, pkg of 100 ea

Synonym(s):

Aluminum crimp seal (without septum), silver aluminum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

silver aluminum

feature

open center

packaging

pkg of 100 ea

manufacturer/tradename

Chromacol 20-ACB

diam.

20 mm

compatibility

for use with 20 mm crimp seal

Looking for similar products? Visit Product Comparison Guide

General description

It is autoclavable, easy sterilization, is used as a closure type for the containers used to contain substances in order to prevent contamination.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service