Skip to Content
Merck
All Photos(1)

Key Documents

26703

Hamilton® syringe, 600 series

62RNR, volume 5 μL, needle size 22s ga, needle L 51 mm (2 in.)

Synonym(s):

Hamilton® Syringe 65RNR, 5UL 22S GA, 2IN, PT#3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
Pricing and availability is not currently available.

material

blunt tip stainless steel needle (point style 3)

needle size

22s ga

description

Microliter

feature

removable needle

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 87943

needle L

51 mm (2 in.)

volume

5 μL

compatibility

for use with Rheodyne

Looking for similar products? Visit Product Comparison Guide

General description

The 600 series syringes [62RNR, volume 5 μL, needle size 22s ga (blunt tip), needle L 51 mm (2 in.)] are ideal for dispensing volumes from 0.25 μL up to 5 μL and is often used in applications that require one handed operation because of the short stroke length and the reinforced plunger.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.
Microliter is a trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Do you have a replacement needle for syringe 26703 that is suitable for use with GC/MS?

    1 answer
    1. The replacement needle 19133-U shares the same dimensions as the needle that comes with syringe 26703, but it features a bevel tip designed for use with GC and GC/MS applications. Another option, part number 19139-U, is a 22s gauge, 51mm long needle with point style 3, similar to the original needle provided with syringe 26703.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service