Skip to Content
Merck
All Photos(1)

Key Documents

20069

Supelco

ORBO 827 LpDNPH Coated Filter, 37mm

pkg of 25 ea

Synonym(s):

ORBO Coated Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900

material

round glass fiber filter

Agency

OSHA (64)
OSHA 81 (2 filters required)
OSHA 85 (3 glass fiber filters required)

product line

ORBO

feature

: no Without cassettes

packaging

pkg of 25 ea

manufacturer/tradename

ORBO 827

technique(s)

active air sampling: suitable

O.D.

37 mm

matrix

LpDNPH coated on glass fiber filter

application(s)

air monitoring
industrial hygiene

compatibility

for analyte group Carbonyls

storage temp.

2-8°C

Legal Information

ORBO is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Flo et al.
Scandinavian journal of clinical and laboratory investigation, 55(8), 715-721 (1995-12-01)
Elevated extracellular cGMP levels have been observed in various clinical conditions, and the analyte has been proposed as a diagnostic marker of cardiovascular as well as malignant diseases. However, the use of extracellular cGMP as a pathophysiological marker requires detailed
A Orbo et al.
Tidsskrift for den Norske laegeforening : tidsskrift for praktisk medicin, ny raekke, 117(7), 972-976 (1997-03-10)
The development of female steroid hormone-related neoplasms such as gynaecologic and mammary cancers is influenced by hormones administered exogenously. Hormonal contraceptives and hormone replacement therapy both affect the risk of developing cancers in the endometrium, cervix, ovarium, and mammary glands.
Bjørn T Moe et al.
Anticancer research, 29(4), 1047-1052 (2009-05-06)
Endometrial hyperplasia is a precursor lesion of endometrial carcinoma. Clinical studies of endometrial hyperplasia have shown that levonorgestrel (LNG) is more therapeutically effective than medroxyprogesterone acetate (MPA). The present pharmacological in vitro study was performed to compare progestin effects on
Anne Ørbo et al.
The Journal of steroid biochemistry and molecular biology, 113(1-2), 139-149 (2009-01-14)
Patients with endometrial hyperplasia representing preliminary stages of endometrial cancer have shown to respond to therapy in 100% of the cases when treated with levonorgestrel-impregnated intrauterine device. Anti-proliferative effect has also been reported after application of an anti-progestin impregnated intrauterine
A Orbo et al.
Gynecologic oncology, 95(1), 82-88 (2004-09-24)
The purpose of the current study was to characterize the role of PTEN in malignant transformation and to evaluate the significance of mutated PTEN exons as prognostic markers in the carcinogenesis of endometrial hyperplasia. A comparison of PTEN mutations as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service