12010AST
Astec® CHIROBIOTIC® T Chiral (5 μm) HPLC Columns
L × I.D. 10 cm × 3 mm, HPLC Column
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52
Recommended Products
Product Name
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 3 mm
material
stainless steel column
Agency
suitable for USP L63
description
HPLC column
product line
Astec®
packaging
pkg of 1 ea
manufacturer/tradename
Astec®
parameter
0-45 °C temperature
241 bar pressure (3500 psi)
technique(s)
HPLC: suitable
LC/MS: suitable
L × I.D.
10 cm × 3 mm
matrix
High-purity silica gel particle platform
fully porous particle
matrix active group
teicoplanin glycopeptide phase
particle size
5 μm
pore size
100 Å
operating pH range
3.8-6.8
separation technique
chiral
Looking for similar products? Visit Product Comparison Guide
General description
CHIROBIOTIC® T and T2 have teicoplanin as the chiral selector. They offer unique selectivity for a number of classes of molecules, specifically underivatized α, β, γ and cyclic amino acids, N-derivatized amino acids, hydroxy-carboxylic acids, acidic compounds including carboxylic acids and phenols, small peptides, neutral aromatic analytes and cyclic aromatic and aliphatic amines. Separations normally obtained on a chiral crown ether or ligand exchange-type CSPs are also possible on the CHIROBIOTIC® T and T2, but in much simpler mobile phases, like water-alcohol. In addition, all of the known beta-blockers (amino alcohols), and dihydrocoumarins have been resolved. CHIROBIOTIC® T and T2 differ in their bonding chemistry and the pore size of the support particle, giving them different selectivity and preparative capacity.
CHIROBIOTIC FAQs
CHIROBIOTIC Reference Bibliography
Chiral Product Literature
- Bonded phase: Teicoplanin
- Operating pH range: 3.8 - 6.8
- Particle diameter: 5, 10 or 16 μm
- Pore size: 100 Å (CHIROBIOTIC® T) or 200 Å (CHIROBIOTIC® T2)
CHIROBIOTIC FAQs
CHIROBIOTIC Reference Bibliography
Chiral Product Literature
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Syntheses of racemic and non-racemic silicon- and germanium-containing a-amino acids of the formula type H2NCH(CH2ElR3)COOH (El=Si, Ge; R=organyl) and incorporation of d-H2NCH(CH2SiMe3)COOH and d-H2NCH(CH2GeMe3)COOH into biologically active decapeptides: a study on C/Si/Ge bioisosterism
Merget, Markus, et al.
Journal of Organometallic Chemistry, 628 (2), 183-194 (2001)
Berit P Jensen et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 865(1-2), 48-54 (2008-03-07)
A stereoselective liquid chromatography-tandem mass spectrometry assay was developed and validated for quantification of S- and R-metoprolol at concentrations of 0.5-50 microg/L in human plasma. Metoprolol was extracted from plasma by liquid-liquid extraction with ethyl acetate (82% recovery). Chromatographic separation
D Abd el-Hady et al.
Journal of pharmaceutical and biomedical analysis, 37(5), 919-925 (2005-05-03)
A simple, sensitive and selective high performance liquid chromatographic method with UV detection for the chiral separation of racemic methotrexate (rac-Mtx) and enantiomeric purity of L-methotrexate in pharmaceutical formulations was developed and validated. The chiral separation was optimized studying both
Knut Gedicke et al.
Journal of chromatography. A, 1162(1), 62-73 (2007-03-03)
The separation of compounds possessing low solubility in the mobile phase could be improved by applying stronger solvents for dissolving the feed. A model system has been investigated with ethanol-water as the mobile phase, Chirobiotic T as the stationary phase
P Jander et al.
Journal of chromatography. A, 919(1), 67-77 (2001-07-19)
The retention behaviour of the enantiomers of underivatized phenylglycine was studied on a Chirobiotic T column packed with amphoteric glycopeptide teicoplanin covalently bonded to the surface of silica gel. The retention and the selectivity of separation of the enantiomers increase
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service