Skip to Content
Merck
All Photos(1)

Documents

Z744127

Round-tip Thirsty-Swabs

pack of 100 ea (swabs per pk)

Synonym(s):

swab kit, swab kits, swabs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111700

packaging

pack of 100 ea (swabs per pk)

manufacturer/tradename

Diversified Biotech SWAB-1000

General description

Round-tip Thirsty-Swabs, 100/pk

Application

Microscopy

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service